Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR761796.2 | - | 9 - 10 | 1355 | 99 | |
CU075779.1 | - | 9 - 10 | 1357 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |