Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203606.2 | NP_988937.1 | 3 - 4 | 268 | 82 | |
GenBank mRNA | BC061397.1 | - | 3 - 4 | 291 | 82 | |
CR761520.2 | - | 3 - 4 | 268 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |