Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhebl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203606.2 | NP_988937.1 | 4 - 5 | 346 | 109 | |
GenBank mRNA | BC061397.1 | - | 4 - 5 | 369 | 109 | |
CR761520.2 | - | 4 - 5 | 346 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |