Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC061362.1 | - | 1 - 2 | 209 | 96 | |
CR848262.2 | - | 1 - 2 | 188 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |