Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012958182.2 | XP_012813636.1 | 3 - 4 | 729 | 72 | |
GenBank mRNA | BC061362.1 | - | 2 - 3 | 376 | 72 | |
CR848262.2 | - | 2 - 3 | 355 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |