Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204041.2 | NP_989372.1 | 4 - 5 | 597 | 96 | |
GenBank mRNA | BC064191.1 | - | 4 - 5 | 594 | 96 | |
CR855594.2 | - | 4 - 5 | 597 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |