Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006887.4 | NP_001006888.1 | 4 - 5 | 391 | 70 | |
GenBank mRNA | BC077687.1 | - | 4 - 5 | 392 | 70 | |
CR926191.2 | - | 334 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |