Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008073.3 | NP_001008074.1 | 4 - 5 | 592 | 118 | |
GenBank mRNA | BC080968.1 | - | 4 - 5 | 517 | 118 | |
CN097570.1 | - | 4 - 5 | 594 | 118 | ||
CR848168.2 | - | 4 - 5 | 431 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |