Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015932.3 | NP_001015932.1 | 1 - 2 | 51 | 88 | |
GenBank mRNA | BC122940.1 | - | 1 - 2 | 51 | 88 | |
CR926375.2 | - | 1 - 2 | 47 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |