Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gadd45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016151.3 | NP_001016151.1 | 3 - 4 | 475 | 74 | |
XM_012961068.2 | XP_012816522.1 | 2 - 3 | 462 | 74 | ||
GenBank mRNA | BC121532.1 | - | 3 - 4 | 475 | 74 | |
BC167898.1 | - | 3 - 4 | 510 | 74 | ||
CR761969.2 | - | 3 - 4 | 501 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |