Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: impad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016215.3 | NP_001016215.2 | 1 - 2 | 389 | 100 | |
GenBank mRNA | BC080464.1 | - | 1 - 2 | 389 | 100 | |
CR926321.2 | - | 1 - 2 | 678 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |