Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc1d7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016332.3 | NP_001016332.2 | 3 - 4 | 598 | 67 | |
XM_012964694.2 | XP_012820148.1 | 2 - 3 | 396 | 67 | ||
GenBank mRNA | BC121213.1 | - | 3 - 4 | 598 | 67 | |
CR761423.2 | - | 2 - 3 | 396 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |