Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016454.3 | NP_001016454.2 | 4 - 5 | 910 | 60 | |
GenBank mRNA | BC124061.1 | - | 4 - 5 | 910 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |