Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nthl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044419.1 | NP_001037884.1 | 1 - 2 | 119 | 85 | |
GenBank mRNA | BC159395.1 | - | 1 - 2 | 167 | 85 | |
CR760124.2 | - | 1 - 2 | 119 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |