Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044434.1 | NP_001037899.1 | 1 - 2 | 101 | 86 | |
GenBank mRNA | CR760562.2 | - | 1 - 2 | 101 | 86 | |
CR926241.1 | - | 1 - 2 | 76 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |