Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc38a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044435.1 | NP_001037900.1 | 5 - 6 | 961 | 83 | |
XM_012961231.2 | XP_012816685.1 | 6 - 7 | 762 | 83 | ||
XM_012961232.2 | XP_012816686.1 | 6 - 7 | 811 | 83 | ||
XM_018093079.1 | XP_017948568.1 | 6 - 7 | 774 | 83 | ||
GenBank mRNA | CR760593.2 | - | 5 - 6 | 961 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |