Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thdl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044446.1 | NP_001037911.1 | 6 - 7 | 821 | 90 | |
XM_018091315.1 | XP_017946804.1 | 4 - 5 | 678 | 90 | ||
GenBank mRNA | CR760814.2 | - | 6 - 7 | 821 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |