Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: krcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045575.1 | NP_001039040.1 | 5 - 6 | 828 | 72 | |
XM_012964252.2 | XP_012819706.1 | 5 - 6 | 898 | 72 | ||
GenBank mRNA | CR848257.2 | - | 5 - 6 | 828 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |