Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: znhit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045581.1 | NP_001039046.1 | 4 - 5 | 283 | 77 | |
GenBank mRNA | BC121593.1 | - | 4 - 5 | 283 | 77 | |
BC157154.1 | - | 4 - 5 | 317 | 77 | ||
CR848407.2 | - | 4 - 5 | 248 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |