Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045595.1 | NP_001039060.1 | 4 - 5 | 408 | 78 | |
GenBank mRNA | BC153678.1 | - | 4 - 5 | 447 | 78 | |
CR848644.1 | - | 4 - 5 | 408 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |