Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgi4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045625.1 | NP_001039090.1 | 6 - 7 | 1121 | 76 | |
GenBank mRNA | CR926431.2 | - | 6 - 7 | 1121 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Xenopus tropicalis Probe spans exons |