Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ipo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045658.1 | NP_001039123.1 | 6 - 7 | 760 | 103 | |
XM_012964360.1 | XP_012819814.1 | 6 - 7 | 760 | 103 | ||
XM_012964366.2 | XP_012819820.1 | 6 - 7 | 760 | 103 | ||
GenBank mRNA | BC168434.1 | - | 6 - 7 | 677 | 103 | |
CR942469.2 | - | 6 - 7 | 760 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |