Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045669.1 | NP_001039134.1 | 2 - 3 | 406 | 66 | |
GenBank mRNA | CR942499.2 | - | 2 - 3 | 406 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |