Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045686.1 | NP_001039151.1 | 2 - 3 | 494 | 80 | |
GenBank mRNA | BC136202.1 | - | 2 - 3 | 442 | 80 | |
CR942568.2 | - | 2 - 3 | 494 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |