Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ogfod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045746.1 | NP_001039211.1 | 3 - 4 | 300 | 86 | |
XM_018094113.1 | XP_017949602.1 | 3 - 4 | 265 | 86 | ||
GenBank mRNA | CR942500.2 | - | 3 - 4 | 300 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |