Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045750.1 | NP_001039215.1 | 4 - 5 | 384 | 92 | |
XM_018093934.1 | XP_017949423.1 | 4 - 5 | 1199 | 92 | ||
GenBank mRNA | BC135657.1 | - | 4 - 5 | 372 | 92 | |
CR942549.2 | - | 4 - 5 | 384 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |