Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045784.1 | NP_001039249.1 | 1410 | 74 | ||
GenBank mRNA | BC168785.1 | - | 11 - 12 | 1100 | 74 | |
CR942792.2 | - | 1410 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |