Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045800.1 | NP_001039265.1 | 2 - 3 | 114 | 80 | |
XM_012959205.2 | XP_012814659.1 | 2 - 3 | 208 | 80 | ||
GenBank mRNA | CR942742.2 | - | 2 - 3 | 114 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |