Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045803.1 | NP_001039268.1 | 7 - 8 | 1174 | 85 | |
XM_012964858.2 | XP_012820312.1 | 8 - 9 | 1385 | 85 | ||
XM_012964859.2 | XP_012820313.1 | 9 - 10 | 1523 | 85 | ||
XM_018094689.1 | XP_017950178.1 | 9 - 10 | 1451 | 85 | ||
GenBank mRNA | BC135988.1 | - | 9 - 10 | 1501 | 85 | |
CR942790.2 | - | 7 - 8 | 1174 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |