Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078709.1 | NP_001072177.1 | 39 - 40 | 2584 | 63 | |
XM_012957183.2 | XP_012812637.1 | 42 - 43 | 2803 | 63 | ||
XM_012957184.2 | XP_012812638.1 | 41 - 42 | 2773 | 63 | ||
XM_012957185.2 | XP_012812639.1 | 41 - 42 | 2770 | 63 | ||
XM_012957188.1 | XP_012812642.1 | 42 - 43 | 2732 | 63 | ||
XM_012957189.2 | XP_012812643.1 | 40 - 41 | 2710 | 63 | ||
XM_012957190.2 | XP_012812644.1 | 40 - 41 | 2682 | 63 | ||
XM_012957191.2 | XP_012812645.1 | 40 - 41 | 2675 | 63 | ||
XM_012957192.2 | XP_012812646.1 | 42 - 43 | 2804 | 63 | ||
XM_012957193.1 | XP_012812647.1 | 42 - 43 | 2805 | 63 | ||
XM_012957194.2 | XP_012812648.1 | 40 - 41 | 2656 | 63 | ||
XM_012957195.2 | XP_012812649.1 | 39 - 40 | 2652 | 63 | ||
XM_012957196.1 | XP_012812650.1 | 42 - 43 | 2804 | 63 | ||
XM_012957197.1 | XP_012812651.1 | 40 - 41 | 2612 | 63 | ||
XM_012957199.2 | XP_012812653.1 | 38 - 39 | 2589 | 63 | ||
XM_012957200.1 | XP_012812654.1 | 42 - 43 | 2805 | 63 | ||
XM_012957204.1 | XP_012812658.1 | 42 - 43 | 2805 | 63 | ||
XM_012957206.1 | XP_012812660.1 | 38 - 39 | 2483 | 63 | ||
GenBank mRNA | DQ523561.1 | - | 39 - 40 | 2584 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Xenopus tropicalis Probe spans exons |