Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Xt03713975_m1
See other ELN1 GE Assays ›
Gene Symbol:
eln1
Gene Name:
tropoelastin 1
Gene Aliases:
eln
Chromosome Location:
Chr.2: 37685140 - 37763602 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID
Xt03713975_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: eln1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001078709.1
NP_001072177.1
10 - 11
639
66
XM_012957183.2
XP_012812637.1
12 - 13
825
66
XM_012957184.2
XP_012812638.1
11 - 12
795
66
XM_012957185.2
XP_012812639.1
12 - 13
825
66
XM_012957188.1
XP_012812642.1
12 - 13
754
66
XM_012957189.2
XP_012812643.1
10 - 11
732
66
XM_012957190.2
XP_012812644.1
10 - 11
704
66
XM_012957191.2
XP_012812645.1
12 - 13
826
66
XM_012957192.2
XP_012812646.1
12 - 13
826
66
XM_012957193.1
XP_012812647.1
12 - 13
827
66
XM_012957194.2
XP_012812648.1
12 - 13
825
66
XM_012957195.2
XP_012812649.1
9 - 10
674
66
XM_012957196.1
XP_012812650.1
12 - 13
826
66
XM_012957197.1
XP_012812651.1
12 - 13
826
66
XM_012957199.2
XP_012812653.1
8 - 9
611
66
XM_012957200.1
XP_012812654.1
12 - 13
827
66
XM_012957201.2
XP_012812655.1
12 - 13
826
66
XM_012957202.1
XP_012812656.1
12 - 13
826
66
XM_012957204.1
XP_012812658.1
12 - 13
827
66
XM_012957205.1
XP_012812659.1
12 - 13
826
66
XM_012957206.1
XP_012812660.1
12 - 13
826
66
XM_012957207.1
XP_012812661.1
12 - 13
827
66
XM_012957208.1
XP_012812662.1
12 - 13
827
66
XM_012957209.1
XP_012812663.1
12 - 13
827
66
XM_012957210.2
XP_012812664.1
12 - 13
827
66
XM_012957211.1
XP_012812665.1
12 - 13
827
66
XM_018091327.1
XP_017946816.1
12 - 13
827
66
GenBank mRNA
DQ523561.1
-
10 - 11
639
66
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Xenopus tropicalis
Probe spans exons
Panther Classification:
Molecular Function -
structural protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0