Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012968587.2 | XP_012824041.1 | 2 - 3 | 794 | 93 | |
XM_012968588.2 | XP_012824042.1 | 2 - 3 | 611 | 93 | ||
GenBank mRNA | BC118726.1 | - | 2 - 3 | 611 | 93 | |
CR761271.2 | - | 2 - 3 | 591 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |