Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078743.1 | NP_001072211.1 | 5 - 6 | 656 | 79 | |
XM_012961289.2 | XP_012816743.1 | 5 - 6 | 678 | 79 | ||
GenBank mRNA | BC118779.1 | - | 5 - 6 | 656 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |