Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078756.1 | NP_001072224.1 | 3 - 4 | 691 | 91 | |
XM_012964975.2 | XP_012820429.1 | 4 - 5 | 855 | 91 | ||
XM_018094299.1 | XP_017949788.1 | 3 - 4 | 751 | 91 | ||
GenBank mRNA | BC118805.1 | - | 3 - 4 | 691 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |