Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ehd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078763.1 | NP_001072231.1 | 3 - 4 | 1049 | 84 | |
XM_012963134.2 | XP_012818588.1 | 4 - 5 | 1248 | 84 | ||
XM_012963135.2 | XP_012818589.1 | 4 - 5 | 1248 | 84 | ||
GenBank mRNA | BC118817.1 | - | 3 - 4 | 1049 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |