Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078786.1 | NP_001072254.1 | 3 - 4 | 626 | 87 | |
XM_012965019.2 | XP_012820473.1 | 3 - 4 | 703 | 87 | ||
XM_012965024.2 | XP_012820478.1 | 3 - 4 | 704 | 87 | ||
GenBank mRNA | BC121219.1 | - | 3 - 4 | 626 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |