Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078791.2 | NP_001072259.2 | 1 - 2 | 433 | 82 | |
GenBank mRNA | BC121245.1 | - | 1 - 2 | 421 | 82 | |
CF152224.1 | - | 1 - 2 | 433 | 82 | ||
CU075823.1 | - | 1 - 2 | 431 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |