Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: txndc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078815.1 | NP_001072283.1 | 670 | 83 | ||
GenBank mRNA | BC121301.1 | - | 670 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis |