Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078817.1 | NP_001072285.1 | 7 - 8 | 1836 | 71 | |
XM_012952548.2 | XP_012808002.1 | 7 - 8 | 2405 | 71 | ||
GenBank mRNA | BC121308.1 | - | 7 - 8 | 1836 | 71 | |
CR761139.2 | - | 4 - 5 | 403 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |