Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078817.1 | NP_001072285.1 | 9 - 10 | 2230 | 97 | |
XM_012952548.2 | XP_012808002.1 | 9 - 10 | 82 | |||
GenBank mRNA | BC121308.1 | - | 9 - 10 | 2230 | 97 | |
CR761139.2 | - | 6 - 7 | 797 | 97 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |