Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qrfpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078827.1 | NP_001072295.1 | 2 - 3 | 1112 | 66 | |
XM_012965103.2 | XP_012820557.1 | 3 - 4 | 738 | 66 | ||
XM_018094374.1 | XP_017949863.1 | 4 - 5 | 731 | 66 | ||
GenBank mRNA | BC121332.1 | - | 2 - 3 | 1112 | 66 | |
CR848550.2 | - | 2 - 3 | 1018 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |