Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078842.3 | NP_001072310.2 | 3 - 4 | 961 | 102 | |
XM_018096302.1 | XP_017951791.1 | 3 - 4 | 549 | 102 | ||
GenBank mRNA | BC121368.1 | - | 3 - 4 | 886 | 102 | |
CR570770.1 | - | 1 - 2 | 66 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |