Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078887.1 | NP_001072355.1 | 3 - 4 | 320 | 62 | |
GenBank mRNA | BC121451.1 | - | 3 - 4 | 320 | 62 | |
BC161546.1 | - | 2 - 3 | 350 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |