Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078949.1 | NP_001072417.1 | 4 - 5 | 429 | 92 | |
GenBank mRNA | BC121647.1 | - | 4 - 5 | 429 | 92 | |
CU025194.1 | - | 4 - 5 | 433 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |