Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078954.2 | NP_001072422.2 | 3 - 4 | 684 | 103 | |
GenBank mRNA | BC121661.1 | - | 3 - 4 | 418 | 103 | |
CR567009.1 | - | 3 - 4 | 684 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |