Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hscb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078954.2 | NP_001072422.2 | 4 - 5 | 831 | 91 | |
XM_018094444.1 | XP_017949933.1 | 3 - 4 | 725 | 91 | ||
GenBank mRNA | BC121661.1 | - | 4 - 5 | 565 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |