Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tesc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078955.1 | NP_001072423.1 | 2 - 3 | 449 | 101 | |
GenBank mRNA | BC121662.1 | - | 2 - 3 | 449 | 101 | |
BC135886.1 | - | 2 - 3 | 300 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |