Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078971.1 | NP_001072439.1 | 5 - 6 | 506 | 77 | |
GenBank mRNA | BC121689.1 | - | 5 - 6 | 506 | 77 | |
BC157239.1 | - | 5 - 6 | 487 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |