Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csrnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078993.1 | NP_001072461.1 | 4 - 5 | 826 | 58 | |
GenBank mRNA | BC121867.1 | - | 4 - 5 | 826 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |