Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079148.1 | NP_001072616.1 | 8 - 9 | 940 | 93 | |
XM_012961357.2 | XP_012816811.1 | 9 - 10 | 1080 | 93 | ||
GenBank mRNA | BC123006.1 | - | 8 - 9 | 940 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |